Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

beta-3 Adrenergic Receptor Polyclonal Antibody, Invitrogen™

Catalog No. PIPA528763
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA528763 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA528763 Supplier Thermo Scientific Supplier No. PA528763

Rabbit Polyclonal Antibody

Recommended positive controls: A431, H1299, HeLa, HepG2, Molt-4, Raji. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

beta 3 Adrenergic Receptor is located mainly in adipose tissue and is involved in the regulation of lipolysis and thermogenesis. Beta adrenergic receptors are involved in the epinephrine and norepinephrine-induced activation of adenylate cyclase through the action of G proteins. beta 3 Adrenergic Receptor expression has been documented in adipose tissues, heart, and in smooth muscle of digestive and urinary tract organs (bladder, colon, small intestine, stomach, ureter). Utilization of alternate promoters and/or 3-prime untranslated regions may result in tissue-specific regulation of the expression of this protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen beta-3 Adrenergic Receptor
Applications Western Blot
Classification Polyclonal
Concentration 0.82 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Gene ADRB3
Gene Accession No. P13945
Gene Alias ADRB; ADRB3; Adrb-3; Adrb3r; adrenergic receptor, beta 3; adrenergic, beta-3-, receptor; adrenoceptor beta 3; B3AR; B3bar; beta 3 adrenoceptor; beta 3-AR; beta-3 adrenergic receptor; Beta-3 adrenoceptor; beta-3 adrenoreceptor; Beta-3 AR; beta3-adrenergic receptor; beta3-Adrenoreceptor; BETA3AR; Beta3-AR; FLJ99960
Gene Symbols ADRB3
Host Species Rabbit
Immunogen Synthetic peptide corresponding to a region within amino acids 344 and 408 of Human beta 3-AR
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 155
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.