Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Beta 3 GABA-A Receptor Monoclonal Antibody (S87-25), QED Bioscience™
SDP

Catalog No. 502278201
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-227-8201 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-227-8201 Supplier Leinco Technologies Inc Supplier No. 56482100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Beta 3 GABA-A Receptor Monoclonal antibody specifically detects Beta 3 GABA-A Receptor in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry, Western Blot

Specifications

Antigen Beta 3 GABA-A Receptor
Applications Immunocytochemistry, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone S87-25
Concentration 0.5-1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.09% sodium azide; pH 7.4
Gene GABRB3
Gene Accession No. P28472, P63079, P63080
Gene Alias A230092K12Rik; AW049585; beta3; cleft palate 1; Cp1; ECA5; GABA(A) receptor subunit beta-3; GABAA receptor beta-3 subunit; GABA-alpha receptor beta-2 subunit; GABR B3; Gabrb3; Gabrb-3; gamma-aminobutyric acid; gamma-aminobutyric acid (GABA) A receptor, beta 3; gamma-aminobutyric acid (GABA) A receptor, subunit beta 3; gamma-aminobutyric acid (GABA-A) receptor, subunit beta 3; gamma-aminobutyric acid A receptor beta 3; Gamma-aminobutyric acid receptor beta 3; gamma-aminobutyric acid receptor subunit beta-3; gamma-aminobutyric acid receptor, subunit beta 3; gamma-aminobutyric acid type A receptor beta3 subunit; gamma-aminobutyric acid type A receptor subunit beta3; LOW QUALITY PROTEIN: gamma-aminobutyric acid receptor subunit beta-3; MGC9051; testis gamma-aminobutyric acid receptor subunit beta 3
Host Species Mouse
Immunogen Fusion protein corresponding to aa 370-433 of mouse Beta 3 GABA-A receptor (accession no. AAB60502).
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14402, 24922, 2562
Target Species Human, Mouse, Rat
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.