Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

beta-3 Tubulin Monoclonal Antibody (2G10-TB3), eBioscience™, Invitrogen™

Mouse Monoclonal Antibody

$176.65 - $1562.00

Specifications

Antigen beta-3 Tubulin
Clone 2G10-TB3
Concentration 0.5 mg/mL
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
14-451-080
Encompass_Preferred
View Documents
Invitrogen™
14451080
25 μg
Each for $176.65

Save 10%

Reg : $196.00

The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Only null left
Edge
Add to Cart
 
14-451-037
Encompass_Preferred
View Documents Promotion Details
Invitrogen™
14451037
2 mg
Each for $1,562.00
Only null left
Add to Cart
 
Description

Description

Description: The 2G10 monoclonal antibody recognizes beta tubulin class III, a member of the large family of tubulins that are components of the microtubules of the cytoskeletal network. Beta tubulin class III expression is exclusively found in neuronal cells of adult mammals. Although expression has been mainly found in neuronal cells, during fetal development, transient expression has been documented in both glial and neuronal precursors localized to the subventricular zone. Identification of beta tubulin class III expression is useful as a structural marker for developing and mature neural cells. Given the high degree of conservation of this protein across species, the 2G10 clone is expected to recognize human, mouse, rat, and bovine beta tubulin class III. Applications Reported: This 2G10 antibody has been reported for use in immunoprecipitation, western blotting, immunohistochemical staining of frozen or formalin-fixed paraffin embedded tissue sections, and immunocytochemistry. Applications Tested: This 2G10 antibody has been tested by immunocytochemistry of fixed and permeabilized cells and can be used at less than or equal to 5 μg/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Purity: Greater than 90%, as determined by SDS-PAGE. Aggregation: Less than 10%, as determined by HPLC. Filtration: 0.2 μm post-manufacturing filtered.

The betaIII-tubulin isoform is present dominantly in cells of neuronal origin and it is one of the earliest markers of neuronal differentiation. It is regarded as a specific probe for the cells of neuronal origin as well as for the tumours originating from these cells. The betaIII-tubulin is most abundant in cells of neuronal origin, but was also detected in Sertoli cells of the testis and transiently in non-neuronal embryonic tissues.
Specifications

Specifications

beta-3 Tubulin
0.5 mg/mL
Unconjugated
Mouse
Human, Mouse, Rat, Bovine
Q13509, Q2T9S0, Q4QRB4, Q9ERD7
IgG2a κ
Affinity chromatography
Antibody
2G10-TB3
Monoclonal
Liquid
RUO
PBS with 0.09% sodium azide; pH 7.2
10381, 22152, 246118, 768070
Primary
4°C
TUBB3
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.