Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ beta-4 Tubulin Monoclonal Antibody (OTI3F1)
GREENER_CHOICE

Catalog No. PIMA525561
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA525561 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA525561 Supplier Invitrogen™ Supplier No. MA525561
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

beta-4 Tubulin Monoclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow

TUBB4A (beta-4 tubulin) encodes a member of the beta tubulin family. Beta tubulins are one of two core protein families (alpha and beta tubulins) that heterodimerize and assemble to form microtubules. Mutations in this gene cause hypomyelinating leukodystrophy-6 and autosomal dominant torsion dystonia-4. Alternate splicing results in multiple transcript variants encoding different isoforms. A pseudogene of this gene is found on chromosome X.
TRUSTED_SUSTAINABILITY

Specifications

Antigen beta-4 Tubulin
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone OTI3F1
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene TUBB4A
Gene Accession No. P04350, Q9D6F9
Gene Alias AI325297; beta-5; class IVa beta-tubulin; dystonia 4, torsion (autosomal dominant); DYT4; hypothetical protein LOC540236; M(beta)4; QtrA-11757; Tubb; TUBB4; Tubb4a; TUBB5; Tubulin 5 beta; tubulin beta; tubulin beta 4A class IVa; tubulin beta-4 chain; Tubulin beta-4A chain; tubulin, beta 4; tubulin, beta 4 class IVa; tubulin, beta 4A class IVa; tubulin, beta, 5; unnamed protein product
Gene Symbols TUBB4A
Host Species Mouse
Immunogen Full length human recombinant protein of TUBB4 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100855815, 10382, 22153, 29213
Target Species Human, Mouse, Rat, Canine, Monkey
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.