Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

beta-Actin Antibody (8H10D10) - BSA Free, Novus Biologicals™
SDP

Catalog No. p-4970237 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP147423SS 0.025 mL
NBP147423 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP147423SS Supplier Novus Biologicals Supplier No. NBP147423SS
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 11 publications

beta-Actin Monoclonal specifically detects beta-Actin in Human, Mouse, Rat, Chinese Hamster, Primate samples. It is validated for Western Blot, Simple Western, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence, Immunoblotting.

Specifications

Antigen beta-Actin
Applications Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence, Immunoblot
Classification Monoclonal
Clone 8H10D10
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000-1:5000, Simple Western 12.5, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunocytochemistry/Immunofluorescence 1:200-1:1000, Immunoblotting reported in scientific literature (PMID 27903531), Flow (Intracellular) 1:200 - 1:400
Formulation PBS with 0.05% Sodium Azide
Gene Accession No. P60709
Gene Alias ACTB, actin, beta, actin, cytoplasmic 1, B-Actin, Beta Actin, beta cytoskeletal actin, Beta-actin, BRWS1, I(2)-Actin, PS1TP5-binding protein 1, PS1TP5BP1
Gene Symbols ACTB
Host Species Mouse
Immunogen This beta-Actin Antibody (8H10D10) was made to a synthetic peptide corresponding to amino-terminal residues of human Beta Actin, conjugated to KLH. [UniProt# P60709]
Molecular Weight of Antigen 42 kDa
Purification Method Ammonium sulfate precipitation
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Cancer, Cell Biology, Cellular Markers, Cytoskeleton Markers, Extracellular Matrix, Loading Controls, Signal Transduction, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 60
Target Species Human, Mouse, Rat, Chinese Hamster, Primate
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Precipitation
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.