Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
beta-Actin Antibody (ACTB/2370), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP32068320UG
Description
beta-Actin Monoclonal antibody specifically detects beta-Actin in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
beta-Actin | |
Monoclonal | |
0.2 mg/ml | |
Immunohistochemistry-Paraffin 1-2 μg/mL | |
ACTB, actin, beta, actin, cytoplasmic 1, B-Actin, Beta Actin, beta cytoskeletal actin, Beta-actin, BRWS1, I(2)-Actin, PS1TP5-binding protein 1, PS1TP5BP1 | |
Recombinant fragment (around aa150-350) of human beta-Actin protein (exact sequence is proprietary) | |
20 μg | |
Angiogenesis, Cancer, Cell Biology, Cellular Markers, Extracellular Matrix, Loading Controls, Signal Transduction, Stem Cell Markers | |
60 | |
Store at 4&drg;C. Do not freeze. | |
IgG |
Immunohistochemistry (Paraffin) | |
ACTB/2370 | |
Unconjugated | |
10mM PBS with 0.05% BSA | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction