Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

beta-Actin Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody has been used in 48 publications

$615.49

Specifications

Antigen beta-Actin
Concentration 1 mg/mL
Dilution Western Blot 1:2000-1:10000, Simple Western 1:2000, Immunohistochemistry 1:2000-1:10000, Immunoprecipitation (Negative), ICC/IF (Negative)
Applications Western Blot, Immunohistochemistry, Immunoprecipitation, Immunocytochemistry
Classification Polyclonal
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB600532
SDP
View Documents
Novus Biologicals
NB600532
0.1 mg
Each of 1 for $615.49
Only null left
Add to Cart
 
Description

Description

beta-Actin Polyclonal specifically detects beta-Actin in Human, Mouse samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunoprecipitation (Negative), ICC/IF (Negative).
Specifications

Specifications

beta-Actin
Western Blot 1:2000-1:10000, Simple Western 1:2000, Immunohistochemistry 1:2000-1:10000, Immunoprecipitation (Negative), ICC/IF (Negative)
Polyclonal
Rabbit
Angiogenesis, Cancer, Cell Biology, Cellular Markers, Cytoskeleton Markers, Extracellular Matrix, Loading Controls, Signal Transduction, Stem Cell Markers
Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
ACTB, actin, beta, actin, cytoplasmic 1, B-Actin, Beta Actin, beta cytoskeletal actin, Beta-actin, BRWS1, I(2)-Actin, PS1TP5-binding protein 1, PS1TP5BP1
ACTB
IgG
Affinity Purified
42 kDa
1 mg/mL
Western Blot, Immunohistochemistry, Immunoprecipitation, Immunocytochemistry
Unconjugated
RUO
Human, Mouse
P60709
60
This beta-Actin Antibody maps to a region corresponding to the N-terminus of human Beta Actin. [UniProt# P60709]
Primary
Store at 4C. Do not freeze.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.