Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

beta-Actin Antibody, Novus Biologicals™
SDP

Catalog No. NB258494 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB258494 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB258494 Supplier Novus Biologicals Supplier No. NBP323319
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

beta-Actin Polyclonal antibody specifically detects beta-Actin in Human, Mouse, Rat, Chicken samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation

Specifications

Antigen beta-Actin
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000, Immunohistochemistry 1:100, Immunocytochemistry/Immunofluorescence 1:50, Immunoprecipitation 1:25
Formulation PBS, 1 mg/ml BSA and 50% glycerol
Gene Alias ACTB, actin, beta, actin, cytoplasmic 1, B-Actin, Beta Actin, beta cytoskeletal actin, Beta-actin, BRWS1, I(2)-Actin, PS1TP5-binding protein 1, PS1TP5BP1
Host Species Rabbit
Immunogen Actin synthetic peptide (coupled to KLH) corresponding to amino acid residues in the N-terminal region of human beta-Actin. This sequence is identical to similar regions in all four alpha-actins, as well as in gamma-actin, and is well conserved in actins from most eukaryotic species. [UniProt# P60709]
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Cancer, Cell Biology, Cellular Markers, Extracellular Matrix, Loading Controls, Signal Transduction, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 60
Target Species Human, Mouse, Rat, Chicken
Content And Storage Store at -20°C.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.