Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

beta Actin Monoclonal Antibody (3H2.E4.E12), Rockland™
SDP

Catalog No. 501990601
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
25 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-0601 25 μL
50-199-0600 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-199-0601 Supplier Rockland Immunochemicals Supplier No. 200301E69S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Store vial at -20°C prior to opening. Aliquot contents and freeze at -20°C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. beta Actin antibody is specific for human and mouse beta Actin protein. Expected to cross-react with a wide range of species due to sequence homology (human, mouse, rat, swine). A BLAST analysis was used to suggest that this antibody would react with beta Actin from a wide range of organisms, including most vertebrates. Cross-reactivity with other sources has not been determined.

Beta actin is one of six actin isoforms which have been identified. Beta actin is a non-muscle cytoskeletal protein in all human cell types and is involved in cell motility, structure, and integrity. There are six different actin isoforms in human. The beta isoform of actin, along with gamma actin, coexist in most cell types as components of the cyto-skeleton. Beta actins are cytoplasmic proteins ubuquitously expressed in all eukaryotic cells. Polymerization of globular actin (G-actin) leads to a structural filament (F-actin) in the form of two stranded helix. Actins are highly conserved proteins that are involved in cell motility, structure and integrity. Because beta actin is ubiquitously expressed in all eukaryotic cells, it is frequently used as a loading control for assays involving protein detection, such as Western blotting. Antibodies to beta-actin provide a specific and useful tool in studying the intracellular distribution of beta-actin and the static and dynamic aspects of the cytoskeleton.

Specifications

Antigen beta Actin
Applications ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 3H2.E4.E12
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2
Gene ACTB
Gene Accession No. P29751, P60709, P60710, P60711, Q6QAQ1
Gene Alias ACT-5; ACTA1; ACTB; actb.L; actb1; actba; actin; actin beta; actin, alpha 1, skeletal muscle; actin, beta; actin, beta 1; actin, beta L homeolog; actin, beta, cytoplasmic; Actin, cytoplasmic 1; Actin, cytoplasmic 1, N-terminally processed; Actx; Atcb; bact; bactin1; bactin1 protein; bactzf; B-ACTZF; beta actin; beta cytoskeletal actin; Beta-actin; Beta-actin-1; BOS_22932; BRWS1; Cap Z; capZ beta; cytoplasmic 1; Cytoplasmic beta-actin; cytoskeletal protein; cytoskeleton actin; E430023M04Rik; gamma actin; gamma non-muscle actin; PS1TP5-binding protein 1; PS1TP5BP1; RCJMB04_4h19; wu:fd18f05; XELAEV_18045052mg
Gene Symbols ACTB
Host Species Mouse
Immunogen Anti-beta Actin (MOUSE) Monoclonal Antibody was produced in mouse by repeated immunizations with beta-Actin peptide near the N-Terminus followed by hybridoma development.
Quantity 25 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100009272, 11461, 414396, 60, 81822
Target Species Human, Mouse, Pig, Rabbit, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a λ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.