Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Beta Actin Monoclonal Antibody (A2-F6), Huabio

Catalog No. 502544773
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-4773 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-254-4773 Supplier HUABIO RESEARCH INC Supplier No. EM21002
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Beta Actin Monoclonal antibody specifically detects Beta Actin in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen Beta Actin
Applications Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone A2-F6
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.2% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Gene ACTB
Gene Accession No. P60709, P60710, P60711
Gene Alias 0610041G09Rik; AA959943; AAT6; ACT; Act4; Act-4; ACT-5; ACTA; acta1; acta1b; ACTA2; Acta-2; ACTA3; actb; actb.L; actb1; actba; ACTC; actc1; Actc-1; actc1b; ACTE; Actg; ACTG1; Actg2; ACTGE; actin; actin alpha 1; actin alpha 1, skeletal muscle; actin alpha 1, skeletal muscle b; actin alpha 2, smooth muscle; actin alpha cardiac; actin alpha cardiac 1; actin alpha cardiac muscle 1; actin alpha cardiac muscle 1b; actin beta; actin gamma 1; actin gamma 2, smooth muscle; actin, alpha 1, skeletal muscle; actin, alpha 1b, skeletal muscle; actin, alpha 2, smooth muscle, aorta; Actin, alpha cardiac muscle 1; Actin, alpha cardiac muscle 1, intermediate form; actin, alpha cardiac muscle 1b; actin, alpha skeletal muscle; Actin, alpha skeletal muscle, intermediate form; actin, alpha, cardiac 1; actin, alpha, cardiac muscle; actin, alpha, cardiac muscle 1; actin, alpha, vascular smooth muscle; actin, aortic smooth muscle; Actin, aortic smooth muscle, intermediate form; actin, beta; actin, beta 1; actin, beta L homeolog; actin, beta, cytoplasmic; actin, cytoplasmic 1; Actin, cytoplasmic 1, N-terminally processed; Actin, cytoplasmic 2; Actin, cytoplasmic 2, N-terminally processed; actin, gamma 1; actin, gamma 2, smooth muscle, enteric; actin, gamma, cytoplasmic 1; actin, gamma-enteric smooth muscle; Actin, gamma-enteric smooth muscle, intermediate form; actin-like protein; Actl; ACTL3; Acts; ACTSA; ACTSG; Actsk-1; Actvs; Actx; AL023024; alpha actin 1; alpha-actin cardiac; alpha-actin-1; Alpha-actin-2; alpha-actin-3; alphac-actin; Alpha-cardiac actin; alphaSMA; alpha-smooth muscle actin; ASD5; ASMA; a-SMA; Bact; Bact; actin; B-actin; bactin1; bactin1 protein; bactzf; B-ACTZF; beta actin; beta cytoskeletal actin; beta-actin; beta-actin FE-3; beta-actin-1; BRWS1; BRWS2; cardiac muscle alpha actin 1; cardiofunk; cell growth-inhibiting gene 46 protein; Cfk; CFTD; CFTD1; CFTDM; CMD1R; CMH11; cytoplasmic 1; cytoplasmic beta-actin; cytoskeletal beta actin; cytoskeletal gamma-actin; cytoskeletal protein; deafness, autosomal dominant 20; deafness, autosomal dominant 26; DFNA20; DFNA26; E430023M04Rik; E51; epididymis luminal protein 176; fa27h01; fb83f06; gamma non-muscle actin; gamma-2-actin; Gamma-actin; gamma-enteric smooth muscle actin; GIG46; HEL-176; hm:zeh0631; I79_002310; I79_013242; I79_019066; LVNC4; MPFD; MYMY5; NEM1; NEM2; NEM3; nemaline myopathy type 3; PS1TP5-binding protein 1; PS1TP5BP1; SHPM; similar to beta actin; skeletal alpha actin; skeletal alpha1 actin; sma; SMalphaA; SMGA; smooth muscle alpha-actin; smooth muscle gamma-actin; vascular smooth muscle alpha-actin; VSCM; wu:fa27h01; wu:fb63d03; wu:fb83f06; wu:fd18f05; XELAEV_18045052mg; zeh0631
Host Species Mouse
Immunogen Synthetic peptide (KLH-coupled) within human Beta-actin aa 1-50.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11461, 60, 81822
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.