Learn More
Beta Actin Mouse, anti-Dog, CoraLite 555, Clone: 2D4H5, Proteintech

Mouse Monoclonal Antibody
Supplier: Proteintech Group Inc CL55566009100UL
Description
The Beta Actin antibody from Proteintech is a mouse monoclonal antibody generated with fusion protein of human Beta Actin. This antibody recognizes human, mouse, rat, hamster, zebrafish, monkey, dog antigen. The Beta Actin antibody has been validated for the following applications: FC (Intra), IF analysis. This antibody is coralite 555.
Specifications
| Beta Actin | |
| 2D4H5 | |
| CoraLite 555 | |
| Mouse | |
| Protein A Purification | |
| Primary | |
| Dog, Hamster, Human, Monkey, Mouse | |
| Liquid |
| Flow Cytometry (Intracellular Staining), Immunofluorescence | |
| 1000 μg/mL | |
| PBS with 50% Glycerol, 0.05% Proclin300, 0.5% BSA, pH 7.3 | |
| Fusion Protein | |
| 100 μL | |
| 60 | |
| Store at -20°C | |
| IgG2b |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.