Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

beta Actin Mouse anti-Human, Mouse, Plant, Porcine, Rat, Zebrafish, Clone: 7D2C10, Proteintech
SDP

Catalog No. 501736190 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-6190 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-6190 Supplier Proteintech Group Inc Supplier No. 600081IG150UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This antibody can recognize all the actins.

The product encoded by this gene belongs to the actin family of proteins, which are highly conserved proteins that play a role in cell motility, structure and integrity. Alpha, beta and gamma actin isoforms have been identified, with alpha actins being a major constituent of the contractile apparatus, while beta and gamma actins are involved in the regulation of cell motility. This actin is an alpha actin that is found in skeletal muscle. Mutations in this gene cause nemaline myopathy type 3, congenital myopathy with excess of thin myofilaments, congenital myopathy with cores, and congenital myopathy with fiber-type disproportion, diseases that lead to muscle fiber defects.

Specifications

Antigen beta Actin
Applications Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 7D2C10
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene ACTA1
Gene Accession No. P60709, P60710, P60711, P62736, P62737, P62738, P63259, P63260, P63261, P63267, P63268, P63269, P68032, P68033, P68035, P68133, P68134, P68136, P68137, Q6QAQ1
Gene Alias ACTB, Actin, actin, beta, Actin, cytoplasmic 1, B actin, Beta actin, F actin
Gene Symbols ACTA1, acta1b, acta2, ACTB, actb1, ACTC1, actc1b, Actg1, ACTG2
Host Species Mouse
Immunogen beta Actin Fusion Protein Ag0297
Purification Method Caprylic Acid Precipitation
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100152267, 100154254, 100520667, 11459, 11461, 11464, 11465, 11468, 11475, 25365, 287876, 29275, 29437, 322509, 397653, 407658, 414396, 57934, 58, 58114, 59, 60, 70, 71, 72, 733615, 81633, 81822
Target Species Human, Mouse, Plant, Pig, Rat, Zebrafish
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgM
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.