Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ beta Actin Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA585291
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA585291 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA585291 Supplier Invitrogen™ Supplier No. PA585291
Add to Cart
Edge
Add to Cart

Rabbit Polyclonal Antibody

Keep as concentrated solution. Predicted reactivity: Mouse (100%), Rat (100%), Japanese Medaka (98%), Xenopus laevis (99%), Dog (100%), Rabbit (99%), Chicken (100%), Sheep (100%), Rhesus Monkey (100%), Chimpanzee (100%), Bovine (100%), Guinea pig (100%). Positive Control: 293T, A431, HeLa, HepG2, Neuro2A, CD830, NIH-3T3, Raw264.7, C2C12, mouse liver, rat liver, PC-12, Rat2, drosophila. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Beta actin is one of six actin isoforms which have been identified. Beta actin is a non-muscle cytoskeletal protein in all human cell types and is involved in cell motility, structure, and integrity. There are six different actin isoforms in human. The beta isoform of actin, along with gamma actin, coexist in most cell types as components of the cyto-skeleton. Beta actins are cytoplasmic proteins ubuquitously expressed in all eukaryotic cells. Polymerization of globular actin (G-actin) leads to a structural filament (F-actin) in the form of two stranded helix. Actins are highly conserved proteins that are involved in cell motility, structure and integrity. Because beta actin is ubiquitously expressed in all eukaryotic cells, it is frequently used as a loading control for assays involving protein detection, such as Western blotting. Antibodies to beta-actin provide a specific and useful tool in studying the intracellular distribution of beta-actin and the static and dynamic aspects of the cytoskeleton.
TRUSTED_SUSTAINABILITY

Specifications

Antigen beta Actin
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.26 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.025% ProClin 300; pH 7
Gene ACTB
Gene Accession No. P60709, P60710, P60711
Gene Alias 0610041G09Rik; AA959943; AAT6; ACT; Act4; Act-4; ACT-5; ACTA; acta1; acta1b; ACTA2; Acta-2; ACTA3; actb; actb.L; actb1; actba; ACTC; actc1; Actc-1; actc1b; ACTE; Actg; ACTG1; Actg2; ACTGE; actin; actin alpha 1; actin alpha 1, skeletal muscle; actin alpha 1, skeletal muscle b; actin alpha 2, smooth muscle; actin alpha cardiac; actin alpha cardiac 1; actin alpha cardiac muscle 1; actin alpha cardiac muscle 1b; actin beta; actin gamma 1; actin gamma 2, smooth muscle; actin, alpha 1, skeletal muscle; actin, alpha 1b, skeletal muscle; actin, alpha 2, smooth muscle, aorta; Actin, alpha cardiac muscle 1; Actin, alpha cardiac muscle 1, intermediate form; actin, alpha cardiac muscle 1b; actin, alpha skeletal muscle; Actin, alpha skeletal muscle, intermediate form; actin, alpha, cardiac 1; actin, alpha, cardiac muscle; actin, alpha, cardiac muscle 1; actin, alpha, vascular smooth muscle; actin, aortic smooth muscle; Actin, aortic smooth muscle, intermediate form; actin, beta; actin, beta 1; actin, beta L homeolog; actin, beta, cytoplasmic; actin, cytoplasmic 1; Actin, cytoplasmic 1, N-terminally processed; Actin, cytoplasmic 2; Actin, cytoplasmic 2, N-terminally processed; actin, gamma 1; actin, gamma 2, smooth muscle, enteric; actin, gamma, cytoplasmic 1; actin, gamma-enteric smooth muscle; Actin, gamma-enteric smooth muscle, intermediate form; actin-like protein; Actl; ACTL3; Acts; ACTSA; ACTSG; Actsk-1; Actvs; Actx; AL023024; alpha actin 1; alpha-actin cardiac; alpha-actin-1; Alpha-actin-2; alpha-actin-3; alphac-actin; Alpha-cardiac actin; alphaSMA; alpha-smooth muscle actin; ASD5; ASMA; a-SMA; Bact; Bact; actin; B-actin; bactin1; bactin1 protein; bactzf; B-ACTZF; beta actin; beta cytoskeletal actin; beta-actin; beta-actin FE-3; beta-actin-1; BRWS1; BRWS2; cardiac muscle alpha actin 1; cardiofunk; cell growth-inhibiting gene 46 protein; Cfk; CFTD; CFTD1; CFTDM; CMD1R; CMH11; cytoplasmic 1; cytoplasmic beta-actin; cytoskeletal beta actin; cytoskeletal gamma-actin; cytoskeletal protein; deafness, autosomal dominant 20; deafness, autosomal dominant 26; DFNA20; DFNA26; E430023M04Rik; E51; epididymis luminal protein 176; fa27h01; fb83f06; gamma non-muscle actin; gamma-2-actin; Gamma-actin; gamma-enteric smooth muscle actin; GIG46; HEL-176; hm:zeh0631; I79_002310; I79_013242; I79_019066; LVNC4; MPFD; MYMY5; NEM1; NEM2; NEM3; nemaline myopathy type 3; PS1TP5-binding protein 1; PS1TP5BP1; SHPM; similar to beta actin; skeletal alpha actin; skeletal alpha1 actin; sma; SMalphaA; SMGA; smooth muscle alpha-actin; smooth muscle gamma-actin; vascular smooth muscle alpha-actin; VSCM; wu:fa27h01; wu:fb63d03; wu:fb83f06; wu:fd18f05; XELAEV_18045052mg; zeh0631
Gene Symbols ACTB
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human beta Actin. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11461, 60, 81822
Target Species Human, Mouse, Rat, Drosophila, Yeast
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.