Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
beta-Actin Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS0061R

Description
Actin exists as a ubiquitous protein involved with filament formation that make up large portions of the cytoskeleton. Actin filaments interact with myosin to assist in muscle contraction as well as aiding in cell motility and cytokinesis. In vertebrates there are three groups of actin isoforms: alpha, beta and gamma. The alpha actins are found in muscle tissues and are a major constituent of the contractile apparatus. The beta and gamma actins co-exists in most cell types as components of the cytoskeleton and as mediators of internal cell motility.Specifications
beta-Actin | |
Polyclonal | |
Unconjugated | |
ACTA2 | |
0610041G09Rik; 5C actin; A; A4V404_DROME; AA959943; a-actin; AAT6; Ac5C; Act; act 5C; ACT1_DROME; Act4; Act-4; Act5; ACT-5; Act5C; Act-5C; Act5C-PA; Act5C-PB; Act5C-PC; Act5C-PD; Act5C-PE; ACTA; Acta1; acta1.S; Acta2; Acta-2; ACTA3; ACTB; actb.L; Actc; Actc1; Actc-1; ACTE; Actg; Actg1; Actg2; ACTGE; Actin; actin 5 C; actin 5C; actin A1; actin alpha 1; actin alpha 1, skeletal muscle; actin alpha 2, smooth muscle; actin alpha cardiac; actin alpha cardiac 1; actin alpha cardiac muscle 1; actin beta; actin gamma 1; actin gamma 2, smooth muscle; actin homolog; actin, alpha 1, skeletal muscle; actin, alpha 1, skeletal muscle S homeolog; actin, alpha 2, smooth muscle, aorta; Actin, alpha cardiac muscle 1; Actin, alpha cardiac muscle 1, intermediate form; Actin, alpha skeletal muscle; actin, alpha skeletal muscle S homeolog; Actin, alpha skeletal muscle, intermediate form; actin, alpha, cardiac 1; actin, alpha, cardiac muscle 1; actin, alpha, vascular smooth muscle; Actin, aortic smooth muscle; Actin, aortic smooth muscle, intermediate form; actin, beta; actin, beta L homeolog; actin, beta, cytoplasmic; Actin, cytoplasmic 1; Actin, cytoplasmic 1, N-terminally processed; Actin, cytoplasmic 2; Actin, cytoplasmic 2, N-terminally processed; Actin, cytoplasmic type 5; actin, cytoplasmic, type 5; actin, gamma 1; actin, gamma 1 propeptide; actin, gamma 2, smooth muscle, enteric; actin, gamma, cytoplasmic 1; Actin, gamma-enteric smooth muscle; Actin, gamma-enteric smooth muscle, intermediate form; Actin/BAP47; Actin5C; Actin-5C; actin-like protein; Actl; ACTL3; Acts; ACTSA; Actsg; Actsk-1; Actvs; Actx; AL023024; alpha 1 skeletal muscle actin; alpha actin; alpha actin 1; alpha actin 2; Alpha-actin; alpha-actin cardiac; Alpha-actin-1; Alpha-actin-2; Alpha-actin-3; alphac-actin; Alpha-cardiac actin; alphaSMA; alpha-sma; alpha-smooth muscle actin; anon-EST:fe2D2; anon-fast-evolving-2D2; ASD5; ASMA; a-SMA; Atcb; Bact; BAP47; beta actin; beta cytoskeletal actin; Beta-actin; beta-actin FE-3; beta-actin/Bap47; BOS_10523; BOS_11389; BOS_19345; BOS_22932; BOS_23286; BOS_23966; BRWS1; BRWS2; cardiac actin; cardiac muscle alpha actin 1; Cell growth-inhibiting gene 46 protein; cellular cytoskeletal beta-actin; CFTD; cftd1; cftdm; CG4027; CG4027 gene product from transcript CG4027-RE; CG4027-PA; CG4027-PB; CG4027-PC; CG4027-PD; CG4027-PE; chrX:5748184..5748304; CMD1R; CMH11; cyt5C; cytoplasmic; cytoplasmic 1; cytoplasmic 2; cytoplasmic actin; Cytoplasmic beta-actin; cytoplasmic type 5; cytoskeletal gamma-actin; cytoskeletal protein; deafness, autosomal dominant 20; deafness, autosomal dominant 26; DFNA20; DFNA26; Dm-actin5C; Dmel\CG4027; Dmel_CG4027; E430023M04Rik; E51; epididymis luminal protein 176; F-actin; G-actin; gamma 1 propeptide; gamma non-muscle actin; Gamma-2-actin; Gamma-actin; gamma-enteric smooth muscle actin; GIG46; HEL-176; I79_002310; I79_013242; I79_019066; l(1)G0009; l(1)G0010; l(1)G0025; l(1)G0079; l(1)G0117; l(1)G0177; l(1)G0245; l(1)G0330; l(1)G0420; l(1)G0486; LOC100009506; LOW QUALITY PROTEIN: actin, aortic smooth muscle; LVNC4; M32055; MPFD; MYMY5; nem1; NEM2; NEM3; nemaline myopathy type 3; PS1TP5-binding protein 1; PS1TP5BP1; RCJMB04_1h13; RCJMB04_4h19; RP5-1068B5.2; SHPM; skeletal alpha actin; sma; SMalphaA; SMGA; smooth muscle alpha-actin; Smooth muscle gamma-actin; T11; type 5; vascular smooth muscle alpha-actin; VSCM; wu:fb63d03; XELAEV_18029136mg; XELAEV_18045052mg | |
Rabbit | |
Protein A | |
RUO | |
58, 59, 60, 70, 71, 72 | |
-20°C | |
Liquid |
Immunofluorescence, Western Blot | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
P60709, P62736, P63261, P63267, P68032, P68133 | |
Acta1, ACTA2, ACTB, ACTC1, ACTG1, ACTG2 | |
MAP conjugated synthetic peptide derived from human beta Actin. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction