Learn More
Beta Actin Rabbit, Clone: RM112, Abnova™
Rabbit monoclonal antibody raised against synthetic peptide of human beta-actin.
Supplier: Abnova Corporation MAB12828
Description
This gene encodes one of six different actin proteins. Actins are highly conserved proteins that are involved in cell motility, structure, and integrity. This actin is a major constituent of the contractile apparatus and one of the two nonmuscle cytoskeletal actins. [provided by RefSeq]
Specifications
Beta Actin | |
Monoclonal | |
Unconjugated | |
ELISA Flow Cytometry Immunocytochemistry (1:200) Immunohistochemistry (1:200) Immunoprecipitation (1:200) Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
PS1TP5BP1 | |
Rabbit | |
Protein A affinity purification | |
RUO | |
60 | |
Human | |
Liquid |
ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot | |
RM112 | |
Rabbit monoclonal antibody raised against synthetic peptide of human beta-actin. | |
In PBS (50% glycerol, 1% BSA, 0.09% sodium azide) | |
ACTB | |
A synthetic peptide corresponding to N-terminus region of human beta-actin. | |
100 μL | |
Primary | |
This antibody reacts to beta-Actin (Actin, cytoplasmic 1). No cross reactivity with other isoforms of Actin. | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.