Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ beta-Actin Recombinant Protein Antigen
SDP

Catalog No. NBP254690X Shop All R&D Systems Products
Change view
Click to view available options
Protein Length:
PKANREKMTQIMFETFNTPAMYVAIQ
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Protein Length
NBP254690X PKANREKMTQIMFETFNTPAMYVAIQ
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP254690X Supplier Novus Biologicals™ Supplier No. NBP254690PEP
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results. Applications: Antibody Competition

A recombinant protein antigen with a N-terminal His6-ABP tag corresponding to human beta-Actin The beta-Actin Recombinant Protein Antigen is derived from E. coli. The beta-Actin Recombinant Protein Antigen has been validated for the following applications: Antibody Competition.

Specifications

Protein Length PKANREKMTQIMFETFNTPAMYVAIQ
Purity >80% by SDS-PAGE and Coomassie blue staining
Common Name ACTB Recombinant Protein Antigen
Content And Storage Store at −20°C. Avoid freeze-thaw cycles.
Formulation PBS and 1M Urea, pH 7.4
For Use With (Application) AC
Gene Alias ACTB, actin, beta, actin, cytoplasmic 1, B-Actin, Beta Actin, beta cytoskeletal actin, Beta-actin, BRWS1, I(2)-Actin, PS1TP5-binding protein 1, PS1TP5BP1
Gene Symbol ACTB
Label Type Unlabeled
Molecular Weight (g/mol) 22 kDa
Product Type Recombinant Protein Antigen
Quantity 0.1 mL
Regulatory Status RUO
Source E.coli
Specific Reactivity This recombinant antigen is only intended to be used as a blocking agent to confirm antibody specificity with the corresponding antibody, catalog number NBP2-53736. It is purified by IMAC chromatography, and the expected concentration is greater than 0.5 mg/ml
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.