Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ beta Adducin Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597179
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597179 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597179 Supplier Invitrogen™ Supplier No. PA597179
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DGEKETAPEE PGSPAKSAPA SPVQSPAKEA ETKSPLVSPS KSLEEGTKKT ETSKAATTEP ETTQPEGVVV NGREEEQTAE EILSKGLSQM TTSADTDVDT SKDKTESVTS G; Positive Samples: U-87MG, 293T, K-562, LO2, HT-1080, A-549, Mouse heart; Cellular Location: Cell membrane, Cytoplasm, Cytoplasmic side, Peripheral membrane protein, cytoskeleton.

Adducins are heteromeric proteins composed of different subunits referred to as adducin alpha, beta and gamma. The three subunits are encoded by distinct genes and belong to a family of membrane skeletal proteins involved in the assembly of spectrin-actin network in erythrocytes and at sites of cell-cell contact in epithelial tissues. While adducins alpha and gamma are ubiquitously expressed, the expression of adducin beta is restricted to brain and hematopoietic tissues. Adducin, originally purified from human erythrocytes, was found to be a heterodimer of adducins alpha and beta. Polymorphisms resulting in amino acid substitutions in these two subunits have been associated with the regulation of blood pressure in an animal model of hypertension. Heterodimers consisting of alpha and gamma subunits have also been described. Structurally, each subunit is comprised of two distinct domains. The amino-terminal region is protease resistant and globular in shape, while the carboxy-terminal region is protease sensitive. The latter contains multiple phosphorylation sites for protein kinase C, the binding site for calmodulin, and is required for association with spectrin and actin. Various adducin beta mRNAs, alternatively spliced at 3'end and/or internally spliced and encoding different isoforms, have been described. The functions of all the different isoforms are not known.
TRUSTED_SUSTAINABILITY

Specifications

Antigen beta Adducin
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.84 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Add2
Gene Accession No. P35612, Q9QYB8
Gene Alias 2900072M03Rik; ADD2; Add97; ADDB; adducin 2; adducin 2 (beta); adducin beta; adducin-63; beta-ADD; Beta-adducin; erythrocyte adducin subunit beta; snoRNA MBII-396
Gene Symbols Add2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 580-690 of human ADD2 (NP_0011719831).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11519, 119
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.