Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Beta-agonist Monoclonal Antibody (1E1)
GREENER_CHOICE

Catalog No. PIMA549617
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA549617 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA549617 Supplier Invitrogen™ Supplier No. MA549617
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Antibody tested in indirect ELISA.

Beta-agonists are a class of drugs that are used in the treatment of various respiratory conditions, such as asthma and chronic obstructive pulmonary disease (COPD). These drugs work by stimulating beta receptors in the lungs, which causes the airways to relax and open up, making it easier to breathe. There are two main types of beta-agonists: short-acting and long-acting. Short-acting beta-agonists are used as rescue medications for acute symptoms, while long-acting beta-agonists are used for maintenance therapy to prevent symptoms from occurring.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Beta-agonist
Applications ELISA, Immunohistochemistry, Lateral Flow, SDS-Page
Classification Monoclonal
Clone 1E1
Concentration 15.9 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Host Species Mouse
Immunogen Conjugate of beta2-AR hapten and KLH.
Purification Method Protein G
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chemical
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.