Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ beta Amylase Polyclonal Antibody, Biotin
GREENER_CHOICE

Catalog No. PIPA127264
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA127264 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA127264 Supplier Invitrogen™ Supplier No. PA127264
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PA1-27264 detects beta Amylase from sweet potato samples.

Beta Amylase is an enzyme found in the seeds of higher plants and in sweet potatoes. It hydrolyses 1,4 alpha D glucosidic linkages in polysaccharides to remove successive maltose units from the non reducing ends of the chains. Used in the structural analysis of starch and glycogen.
TRUSTED_SUSTAINABILITY

Specifications

Antigen beta Amylase
Applications Dot Blot, ELISA, Western Blot
Classification Polyclonal
Concentration 10 mg/mL
Conjugate Biotin
Formulation PBS with 1% BSA and 0.01% sodium azide; pH 7.2
Gene Alias ß-amylase
Host Species Rabbit
Immunogen Beta Amylase (Sweet Potato).
Purification Method IgG fraction
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Plant
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.