Learn More
Invitrogen™ beta Amyloid (1-40) Polyclonal Antibody, Biotin

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ 443489
Description
This beta Amyloid (1-40) antibody (44-3489) is purified from rabbit serum by epitope-specific affinity chromatography. Any reactivity towards both full length Abeta 42 and A beta 43 protein has been eliminated through a series of preabsorption steps. This antibody shows no significant cross-reactivity against Abeta 42 or Abeta 43.
Beta-amyloid is a 40-43 amino acid peptide cleaved from amyloid precursor protein, and a putative gamma-secretase. Amyloid beta (1-40) together with Amyloid beta (1-42) are two major C-terminal variants of the amyloid beta protein, constituting the majority of amyloid beta proteins. Both the amyloid beta (40) and amyloid beta (42) alloforms are major components of the extracellular plaques found in Alzheimer's disease brain tissue.
Specifications
| beta Amyloid (1-40) | |
| Polyclonal | |
| Biotin | |
| APP | |
| A beta 1 40; A ß 1 40; A4; AAA; AB40; Abeta; Abeta40; Abeta42; ABPP; AD1; Adap; AG; AICD-50; AICD-57; AICD-59; AID(50); AID(57); AID(59); Alpha-CTF; Alpha-secretase C-terminal fragment; Alzheimer disease amyloid A4 protein homolog; alzheimer disease amyloid protein; Amyloid; Amyloid b; Amyloid beta; amyloid beta (A4) precursor protein; Amyloid beta 1 40; amyloid beta A4 protein; amyloid beta precursor protein; Amyloid intracellular domain 50; Amyloid intracellular domain 57; Amyloid intracellular domain 59; Amyloid precursor protein; Amyloid ß; Amyloid-beta A4 protein; Amyloid-beta precursor protein; Amyloid-beta protein 40; Amyloid-beta protein 42; Amyloidogenic glycoprotein; App; APP-C57; APP-C59; APP-C99; APPI; appican; Aß 1 40; B APP40; BAPP40; beta-amyloid peptide; beta-amyloid peptide(1-40); beta-amyloid peptide(1-42); beta-amyloid precursor protein; betaApp; BetaAPP40; Beta-APP40; Beta-APP42; Beta-CTF; Beta-secretase C-terminal fragment; C31; C80; C83; C99; Cerebral vascular amyloid peptide; CTFgamma; CVAP; E030013M08Rik; Gamma-CTF(50); Gamma-CTF(57); Gamma-CTF(59); Gamma-secretase C-terminal fragment 50; Gamma-secretase C-terminal fragment 57; Gamma-secretase C-terminal fragment 59; N-APP; P3(40); P3(42); peptidase nexin-II; PN2; PN-II; PreA4; protease nexin II; protease nexin-II; S-APP-alpha; S-APP-beta; Soluble APP-alpha; Soluble APP-beta; ß APP40; ßAPP40; testicular tissue protein Li 2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 351 | |
| -80°C | |
| Liquid |
| Dot Blot, Functional Assay, Radioimmune Assays (RIA) | |
| 1 mg/mL | |
| PBS with 0.1% sodium azide; pH 7.2 | |
| P05067 | |
| APP | |
| 1-40 of Amyloid Beta. | |
| 25 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.