Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

beta-Amyloid 1-42 Mouse anti-Human, Clone: 2C2G5, GenScript™

Catalog No. 50273684
Encompass_Preferred
Change view
Click to view available options
Quantity:
40 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-273-684 40 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-273-684 Supplier Genscript Corporation Supplier No. A0156740
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Reconstitute 40 μg lyophilized powder with 80 μl of deionized water (or equivalent) to antibody concentration of 0.5 mg/mL. A number of mutations, identified in the gene encoding the β-amyloid precursor protein (βAPP), have been linked to early-onset Familial Alzheimers Disease. βAPP is cleaved sequentially by the proteolytic enzymes β-secretase and γ-secretase to produce β-amyloid (Aβ) peptides with the Aβ1-42(43) and the Aβ1-40 forms being the most prevalent. Secreted Aβ peptides can bind to scavenger receptors and the receptor for advanced glycation end products. Aβ peptides are degraded either via a reuptake mechanism followed by endosomal degradation or by an extracellular insulin-degrading enzyme. Extracellular accumulation of Aβ leads to formation of aggregates, fibrils, and eventually amyloid deposits called neuritic plaques, a hallmark of Alzheimer's disease. β-amyloid antibodies and peptides have been developed as tools for elucidating the biology of Alzheimers disease.GenScript Human β-Amyloid 1-42 Antibody (2C2G5), mAb, Mouse is produced from the hybridoma resulting from fusion of Sp2/0 myeloma and B-lymphocytes obtained from mouse immunized with a synthetic peptide corresponding to amino acids 36-42 of human β-Amyloid conjugated to KLH (Swiss Prot: P05067).

beta-Amyloid 1-42 Monoclonal antibody specifically detects beta-Amyloid 1-42 in Human samples. It is validated for Dot Blot, ELISA, Immunohistochemistry (Paraffin)

Specifications

Antigen beta-Amyloid 1-42
Applications Dot Blot, ELISA, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 2C2G5
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide; pH 7.4
Gene APP
Gene Accession No. P05067
Gene Alias A beta 1 42; A ß 1 42; A4; AAA; AB42; Abeta; Abeta40; Abeta42; ABPP; AD1; Adap; AG; AICD-50; AICD-57; AICD-59; AID(50); AID(57); AID(59); Alpha-CTF; Alpha-secretase C-terminal fragment; Alzheimer disease amyloid A4 protein homolog; alzheimer disease amyloid protein; Amyloid; amyloid A4; Amyloid b; Amyloid beta; amyloid beta (A4) precursor protein; Amyloid beta 1 42; amyloid beta A4 protein; amyloid beta precursor protein; Amyloid intracellular domain 50; Amyloid intracellular domain 57; Amyloid intracellular domain 59; Amyloid precursor protein; Amyloid ß; Amyloid-beta A4 protein; Amyloid-beta precursor protein; Amyloid-beta protein 40; Amyloid-beta protein 42; Amyloidogenic glycoprotein; APP; APP-C57; APP-C59; APP-C99; APPI; appican; Aß 1 42; B APP42; BAPP42; beta-amyloid peptide; beta-amyloid peptide(1-40); beta-amyloid peptide(1-42); beta-amyloid precursor protein; betaApp; Beta-APP40; BetaAPP42; Beta-APP42; Beta-CTF; Beta-secretase C-terminal fragment; C31; C80; C83; C99; Cerebral vascular amyloid peptide; CTFgamma; Cvap; E030013M08Rik; Gamma-CTF(50); Gamma-CTF(57); Gamma-CTF(59); Gamma-secretase C-terminal fragment 50; Gamma-secretase C-terminal fragment 57; Gamma-secretase C-terminal fragment 59; N-APP; P3(40); P3(42); peptidase nexin-II; PN2; PN-II; PreA4; protease nexin II; protease nexin-II; S-APP-alpha; S-APP-beta; Soluble APP-alpha; Soluble APP-beta; testicular tissue protein Li 2; ß APP42; ßAPP42
Host Species Mouse
Immunogen KLH conjugated synthetic peptide derived from human beta-Amyloid 1-42
Purification Method Affinity chromatography
Quantity 40 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 351
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.