Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
beta Amyloid Antibody (MOAB-2), Allophycocyanin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP213075APC
Description
beta Amyloid Monoclonal antibody specifically detects beta Amyloid in Human, Mouse, Rat, Bacteria, Monkey samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoblot, Dot Blot, Immunohistochemistry Free-FloatingSpecifications
beta Amyloid | |
Monoclonal | |
APC | |
PBS | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Human, Mouse, Rat, Bacteria, Monkey | |
Purified |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunoprecipitation | |
MOAB-2 | |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunoblotting, Dot Blot, Immunohistochemistry Free-Floating | |
Abeta, Amyloid beta, beta-amyloid peptide | |
This beta Amyloid antibody was developed against recombinant human beta Amyloid 42. | |
0.1 mL | |
Alzheimers Research, Apoptosis, Cardiovascular Biology, Cell Cycle and Replication, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Phospho Specific, Signal Transduction | |
351 | |
Store at 4C in the dark. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction