Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
beta Amyloid Antibody (MOAB-2) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP280594
Description
beta Amyloid Monoclonal specifically detects beta Amyloid in Human, Mouse, Rat, Monkey samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunoblotting, Dot Blot, Immunohistochemistry Free-Floating, Immunocytochemistry.Specifications
beta Amyloid | |
Monoclonal | |
1 mg/mL | |
Western Blot : 1:1000-1:5000, ELISA : 1:100-1:1000, Immunohistochemistry : 1:40-1:400, Immunocytochemistry/Immunofluorescence : 1:200-1:500, Immunoprecipitation : 1:200-1:1000, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunoblotting reported in scientific literature (PMID 28314768), Dot Blot reported in scientific literature (PMID 22423893), Immunohistochemistry Free-Floating reported in scientific literature (PMID 25747037), Immunocytochemistry | |
Abeta, Amyloid beta, beta-amyloid peptide | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Human, mouse and rat. Monkey reactivity reported in scientific literature (PMID: 29241829). | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG2b |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoblot, Dot Blot, Immunohistochemistry (Free Floating) | |
MOAB-2 | |
Unconjugated | |
PBS | |
APP | |
This beta Amyloid antibody was developed against recombinant human beta Amyloid 42. | |
0.1 mL | |
Alzheimers Research, Apoptosis, Cardiovascular Biology, Cell Cycle and Replication, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Phospho Specific, Signal Transduction | |
351 | |
Human, Mouse, Rat, Monkey | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction