Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

beta Amyloid Mouse anti-Human, Porcine, Clone: 5C2A1, Proteintech
SDP

Catalog No. 501736429 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-6429 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-6429 Supplier Proteintech Group Inc Supplier No. 603421IG150UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Amyloid beta peptide (Abeta) is the major constituent of amyloid plaques in the brains of individuals afflicted with Alzheimer′s disease. This peptide is generated from the beta-amyloid precursor protein (beta APP) in a two-step process. The first step involves cleavage of the extracellular, amino-terminal domain of beta APP. Protein cleavage is performed by an aspartyl protease termed beta-secretase (BACE). This enzyme is synthesized as a propeptide that must be modified to the mature and active form by the prohormone convertase, furin. Beta APP cleavage by the mature form of BACE results in the cellular secretion of a segment of beta APP and a membrane-bound remt. This remt is then processed by another protease termed gamma-secretase. Gamma-secretase cleaves an intra-membrane site in the carboxyl-terminal domain of beta APP, thus generating the amyloid beta peptide. Gamma-secretase is believed to be a multi-subunit complex containing presenilin-1 and 2 as central components. Found associated with the presenilins is the transmembrane glycoprotein nicastrin. Nicastrin has been found to bind to the carboxyl-terminus of betaAPP and helps to modulate the production of the amyloid beta peptide Also found in the neurofibrillary lesions in Alzheimer′s disease is the protein termed Tau. Tau is a neuronal microtubule-associated protein found predomitly on axons. The function of tau is to promote tubulin polymerization and stabilize microtubules. Tau, in its hyperphosphorylated form, is the major component of paired helical filaments (PHF), which is the building block of neurofibrillary lesions in Alzheimer′s disease brain.

Specifications

Antigen beta Amyloid
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 5C2A1
Concentration 1.12 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene APP
Gene Accession No. P05067, P79307
Gene Alias A4, AAA, ABETA, ABPP, AD1, AICD 50, AICD 57, AICD 59, AID(50), AID(57), AID(59), Amyloid beta A4 protein, amyloid P, APP, APPI, beta Amyloid, CTFgamma, CVAP, Gamma CTF(50) C31, PN II, PN2, PreA4, Protease nexin II, S APP alpha Soluble APP beta
Gene Symbols APP
Host Species Mouse
Immunogen beta Amyloid Fusion Protein Ag0769
Purification Method Protein A
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 351, 397663
Target Species Human, Pig
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.