Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
beta Amyloid, Pyro Glu11 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP144070
Description
beta Amyloid Polyclonal specifically detects beta Amyloid in Human, Drosophila, Primate samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoassay.Specifications
beta Amyloid (Pyro Glu11) | |
Polyclonal | |
Unconjugated | |
0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 30% Glycerol with No Preservative | |
Abeta, Amyloid beta, beta-amyloid peptide | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
This antibody is specific to the N-terminus 11-pyro E start point. | |
Human, Drosophila, Primate | |
IgG |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoassay | |
LYOPH | |
Western Blot 0.2 ug/ml, ELISA 1:20000-1:60000, Immunohistochemistry 1:100-1:500, Immunocytochemistry/Immunofluorescence 1:100-1:500, Immunoassay | |
P05067 | |
APP | |
beta Amyloid [Pyro Glu11] Antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding to the N-terminus 11-pyro E start point of human beta Amyloid. (Uniprot: P05067) | |
0.05 mg | |
Alzheimers Research, Apoptosis, Cardiovascular Biology, Cell Cycle and Replication, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Signal Transduction | |
351 | |
Reconstitute with 50 ul deionized water (or equivalent) | |
Store at -20°C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction