Learn More
Invitrogen™ beta-Arrestin 1,2 Polyclonal Antibody

Description
PA1-730 detects recombinant rat and human beta-arrestin and beta-arrestin2. This antibody does not detect visual or cone arrestin. PA1-730 has been successfully used in Western blot, Immunohistochemistry (paraffin) and immunoprecipitation procedures. By Western blot, this antibody detects ~49 kDa and ~47 kDa proteins representing recombinant beta-arrestin and beta-arrestin2, respectively. The PA1-730 immunizing peptide corresponds to amino acid residues 384-397 from human beta-arrestin2. This peptide (Cat. # PEP-156) is available for use in neutralization and control experiments.
Specifications
Specifications
| Antigen | beta-Arrestin 1,2 |
| Applications | Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1 mg/mL BSA and 0.05% sodium azide |
| Gene | ARRB2 |
| Gene Accession No. | P29066, P29067, P32121, P49407 |
| Gene Alias | ARB1; ARB2; ARR1; ARR2; Arrb1; Arrb2; arrestin 2; arrestin 3; arrestin beta 1; arrestin beta 2; arrestin beta-1; Arrestin beta-2; arrestin, beta 1; arrestin, beta 2; BARR2; BARRES; Beta-arrestin-1; beta-arrestin-2; Non-visual arrestin-2; Non-visual arrestin-3 |
| Gene Symbols | ARRB1, ARRB2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.