Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ beta Arrestin 1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA586418
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA586418 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA586418 Supplier Invitrogen™ Supplier No. PA586418
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Purity is > 95% by SDS-PAGE.

Beta Arrestin 1 is a member of a family of proteins widely expressed but especially abundant in the central nervous system. Serving as an adaptor or scaffold molecule, beta Arrestin 1 is essential for mitogenic signaling and mediates agonist-dependent desensitization and internalization of Gprotein-coupled receptors (GPCRs, e.g., beta 2-adrenergic receptor). After binding to their ligand and interacting with heterotrimeric G proteins, GPCRs are phosphorylated by G-protein receptor kinases (GRKs) on serine residues. Beta Arrestin 1 in the cytosol is phosphorylated by ERK1 and 2 on serine412 in a negative feedback mechanism and binds to the phosphorylated receptors at the plasma membrane. Serine 412 is then dephosphorylated and the GPCRs are internalized, leading to activation of the Ras, Raf, ERK1 and 2 signaling pathway.
TRUSTED_SUSTAINABILITY

Specifications

Antigen beta Arrestin 1
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene ARRB1
Gene Accession No. P29066, P49407, Q8BWG8
Gene Alias 1200006I17Rik; ARB1; ARR1; ARRB1; ARRB1 protein; arrdc1; arrestin; arrestin 2; arrestin beta 1; arrestin beta-1; arrestin, beta 1; arrestin-2; AW208571; BARRES; beta-arrestin; beta-arrestin1; beta-arrestin-1; G430100A01Rik; Non-visual arrestin-2
Gene Symbols ARRB1
Host Species Rabbit
Immunogen Synthetic peptide, corresponding to amino acids 360-410 of Human Arrestin-beta-1.
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 109689, 25387, 408
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.