Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

beta-Arrestin-2 Mouse anti-Human, Rat, Clone: 5E12.1, MilliporeSigma™

Catalog No. MABN1580MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABN1580MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABN1580MI Supplier MilliporeSigma Supplier No. MABN1580
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-beta-Arrestin-2 Antibody, clone 5E12.1 is an antibody against beta-Arrestin-2 for use in Western Blotting, Immunohistochemistry (Paraffin).
TRUSTED_SUSTAINABILITY

Specifications

Antigen beta-Arrestin-2
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 5E12.1
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Gene Symbols ARRB2,ARB2,ARR2,BARR2
Host Species Mouse
Immunogen KLH-conjugated linear peptide corresponding to a sequence in the C-terminal region of human beta-Arrestin-2.
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) NP_001244257
Test Specificity Expected to react with spliced isoforms 1, 4, and 5 reported by UniProt (P32121). Reactivity toward spliced isoforms 2 & 3 has not been determined.
Target Species Human, Rat
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.