Learn More
Invitrogen™ beta-Arrestin 2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA588224
Description
Immunogen sequence: NLASSTIVKE GANKEVLGIL VSYRVKVKLV VSRGGDVSVE LPFVLMHPKP HDHIPLPRPQ SAAPETDVPV DTNLIEFDTN YATDDDIVFE DFARLRLKGM K; Positive Samples: U-251MG, A-549; Cellular Location: Cell membrane, Cytoplasm, Cytoplasmic vesicle, Membrane, Nucleus, clathrin-coated pit.
Beta-arrestin-2, or ARRB2, is an adaptor protein belonging to the arrestin family with an N- arrestin domain and a C- arrestin domain. ARRB2 is a regulatory protein involved in heterotrimeric G protein-coupled receptor desensitization and is known to regulate beta2-adrenergic receptor (beta2AR) function by enhancing beta2AR mediated nuclear translocation of ERK. ARRB2 binds to phosphorylated beta2ARs, thereby causing a significant impairment of their capacity to activate G(S) proteins. Along with AIP4, ARRB2 acts as an endosomal sorting molecule that mediates CXCR4 entry into a degradative pathway. It may also be involved in hormone-specific desensitization of TSH receptors. ARRB2 is a Ca2+ binding protein of the retinal outer segments and binds to P-rhodopsin and is also involved in synaptic transmission in photoreceptor cells. It is widely expressed in most of the tissues, especially neuronal tissues and spleen.
Specifications
beta-Arrestin 2 | |
Polyclonal | |
Unconjugated | |
ARRB2 | |
AI326910; ARB2; ARR2; ARRB2; arrestin 3; arrestin beta 2; arrestin beta-2; arrestin, beta 2; arrestin-3; AW122872; BARR2; BARRES; beta arr2; beta-arrestin 2 long isoform; beta-arrestin 2 short isoform; beta-arrestin-2; Non-visual arrestin-3 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
216869, 25388, 409 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.89 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P29067, P32121, Q91YI4 | |
ARRB2 | |
A synthetic peptide corresponding to a sequence within amino acids 300-400 of human ARRB2 (NP_0043041). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.