Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Beta Caryophyllene, Natural Isolate, Ricca Chemical
SDP

Catalog No. 01184982
Click to view available options
Quantity:
20 g
100 g
750 g

Beta Caryophyllene, Natural Isolate is a natural isolate food additive.

  • All Natural
  • Food Grade
  • FSSC 22000 and ISO 9001 Accreditated Facility
  • Additional Food Safety Testing Included (Pesticide, Residual Solvents, Heavy Metals)

Specifications

Chemical Name or Material Beta Caryophyllene
Packaging Amber Glass
Certificate of Analysis Yes
CAS 87-44-5
Odor Woody, spicy, clove-like
Assay Percent Range 80% to 100%
Physical Form Liquid
Molecular Formula C15H24
Color Colorless to Light Yellow
Type Food Additive Standard
Molecular Weight (g/mol) 204.36
Quantity 750 g
Grade Food Grade
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.