Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
beta Casein Antibody (F20.14), Alexa Fluor™ 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB1002720AF350
Description
beta Casein Monoclonal specifically detects beta Casein in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
beta Casein | |
Monoclonal | |
Alexa Fluor 350 | |
50 mM sodium borate with 0.05% sodium azide | |
CSN2 | |
Purified casein and a-lactalbumin. | |
0.1 mL | |
1447.0 | |
Human | |
IgG1 |
Western Blot, Immunohistochemistry | |
F20.14 | |
Western Blot, Immunohistochemistry | |
beta-casein, CASB, casein beta | |
Mouse | |
Protein A purified | |
Primary | |
Does not cross react with alpha-lactalbumin. | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction