Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
beta Casein Antibody (F20.14), Alexa Fluor™ 594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB1002720AF594
Description
beta Casein Monoclonal specifically detects beta Casein in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
| beta Casein | |
| Monoclonal | |
| Alexa Fluor 594 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| CSN2 | |
| Purified casein and a-lactalbumin. | |
| 0.1 mL | |
| 1447.0 | |
| Human | |
| IgG1 |
| Western Blot, Immunohistochemistry | |
| F20.14 | |
| Western Blot, Immunohistochemistry | |
| beta-casein, CASB, casein beta | |
| Mouse | |
| Protein A purified | |
| Primary | |
| Does not cross react with alpha-lactalbumin. | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction