Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
beta Casein Antibody (F20.14), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB1002720F
Description
beta Casein Monoclonal specifically detects beta Casein in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Frozen.Specifications
beta Casein | |
Monoclonal | |
FITC | |
PBS with 0.05% Sodium Azide | |
CSN2 | |
Purified casein and a-lactalbumin. | |
0.1 mL | |
Primary | |
Does not cross react with alpha-lactalbumin. | |
Store at 4C in the dark. | |
IgG1 |
Immunohistochemistry, Immunohistochemistry (Frozen) | |
F20.14 | |
Immunohistochemistry | |
beta-casein, CASB, casein beta | |
Mouse | |
Protein A purified | |
RUO | |
1447 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction