Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
beta Casein Antibody (SR1391), Novus Biologicals™

Rabbit Monoclonal Antibody
$309.50 - $423.00
Specifications
Antigen | beta Casein |
---|---|
Clone | SR1391 |
Dilution | Western Blot 1:500-1:1000, Immunoprecipitation 1:50 |
Applications | Western Blot, Immunoprecipitation |
Classification | Monoclonal |
Description
beta Casein Monoclonal antibody specifically detects beta Casein in Human samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
beta Casein | |
Western Blot 1:500-1:1000, Immunoprecipitation 1:50 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
beta-casein, CASB, casein beta | |
A synthesized peptide derived from human beta Casein (Uniprot #: P05814) | |
Primary | |
Store at -20° C. Avoid freeze/thaw cycles. |
SR1391 | |
Western Blot, Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Cancer, Endocrinology, Signal Transduction | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
1447 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title