Learn More
Invitrogen™ Beta-casein Polyclonal Antibody
Description
Beta-casein Polyclonal Antibody for Western Blot, IHC (P)
Specifications
Specifications
| Antigen | Beta-casein |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 |
| Gene | CSN2 |
| Gene Accession No. | P02665, P05814, P10598 |
| Gene Alias | beta casein; beta-caesin; beta-casein; beta-casein A3; beta-casein precursor; CASB; casein beta; CSN2; kappa-casein; Unknown (protein for MGC:133765); unnamed protein product |
| Gene Symbols | CSN2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.