Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Beta-casein Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5109599
Description
Immunogen sequence: MKVLILACLV ALALARETIE SLSSSEESIT EYKQKVEKVK HEDQQQGEDE HQDKIYPSFQ PQPLIYPFVE PIPYGFLPQN ILPLAQPAVV LPVPQPEIME.
This gene is a member of the beta casein family. There are two types of casein protein, beta (encoded by this gene) and kappa, both of which are secreted in human milk. Beta casein is the principal protein in human milk and the primary source of essential amino acids for a suckling infant. Beta and kappa casein proteins acting together form spherical micelles which bind within them important dietary minerals, such as calcium and phosphorous. Alternative splicing results in multiple transcript variants.
Specifications
Beta-casein | |
Polyclonal | |
Unconjugated | |
CSN2 | |
beta casein; beta-caesin; beta-casein; beta-casein A3; beta-casein precursor; CASB; casein beta; CSN2; kappa-casein; Unknown (protein for MGC:133765); unnamed protein product | |
Rabbit | |
Affinity chromatography | |
RUO | |
1447 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
3.14 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
P05814 | |
CSN2 | |
A synthetic peptide corresponding to a sequence within amino acids 1-100 of human CSN2 (NP_001882.1). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction