Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
beta casein Rabbit anti-Bovine∣Goat, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS10032R

Description
Important role in determination of the surface properties of the casein micelles. Casoparan acts as a macrophage activator, increasing the phagocytic activity of macrophages and peroxide release from macrophages. It also acts as a bradykinin-potentiating peptide. Casohypotensin acts as a bradykinin-potentiating peptide. Induces hypotension in rats. Acts as a strong competitive inhibitor of endo-oligopeptidase A. Antioxidant peptide has antioxidant activity.Specifications
beta casein | |
Polyclonal | |
Unconjugated | |
CSN2 | |
CSN2 | |
KLH conjugated synthetic peptide derived from bovine Beta-casein. | |
100 μL | |
Primary | |
Bovine, Goat | |
Antibody | |
IgG |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
P02666, P33048 | |
Rabbit | |
Protein A | |
RUO | |
100860784, 281099 | |
-20°C | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction