Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

beta casein Rabbit anti-Ovine, Polyclonal, Bioss
SDP

Catalog No. 501980292
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-0292 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-0292 Supplier Bioss Supplier No. BS0466R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Important role in determination of the surface properties of the casein micelles. Casoparan acts as a macrophage activator, increasing the phagocytic activity of macrophages and peroxide release from macrophages. It also acts as a bradykinin-potentiating peptide. Casohypotensin acts as a bradykinin-potentiating peptide. Induces hypotension in rats. Acts as a strong competitive inhibitor of endo-oligopeptidase A. Antioxidant peptide has antioxidant activity.

Specifications

Antigen beta casein
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene CSN2
Gene Accession No. P11839
Gene Symbols CSN2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from sheep Beta-casein .
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 443391
Target Species Ovine
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.