Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

beta-Catenin Antibody (9F2) - Azide and BSA Free, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

$690.42

Specifications

Antigen beta-Catenin
Clone 9F2
Concentration 1 mg/mL
Dilution Western Blot 0.5 - 1 ug/ml, Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 1 - 2 ug/ml, CyTOF-ready
Applications Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, CyTOF
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB254495100
SDP
View Documents
Novus Biologicals
NBP254495100UG
100 μg
Each for $690.42
Only null left
Add to Cart
 
NB254495200
SDP
View Documents
Novus Biologicals
NBP254495200UG
200 μg N/A N/A N/A
Description

Description

beta-Catenin Monoclonal specifically detects beta-Catenin in Human, Bovine, Canine, Chicken samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, CyTOF-ready.
Specifications

Specifications

beta-Catenin
1 mg/mL
Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, CyTOF
Unconjugated
Mouse
Cancer, Cellular Markers, Extracellular Matrix, Mucosal Immunology, Neuroscience, Phospho Specific, Stem Cell Signaling Pathway, Wnt Signaling Pathway
PBS with No Preservative
1499
Recombinant chicken Beta-catenin
Primary
Store at -20 to -80C. Avoid freeze-thaw cycles.
9F2
Western Blot 0.5 - 1 ug/ml, Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 1 - 2 ug/ml, CyTOF-ready
Monoclonal
Purified
RUO
Human, Bovine, Canine, Chicken
beta 1 (88kD), beta-catenin, catenin (cadherin-associated protein), beta 1, 88kDa, catenin beta-1, CTNNB, DKFZp686D02253, FLJ25606, FLJ37923
CTNNB1
IgG1 κ
Protein A or G purified
92 kDa
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.