Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
beta-Catenin Antibody (CTNNB1/1509) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$707.00
Specifications
| Antigen | beta-Catenin |
|---|---|
| Clone | CTNNB1/1509 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 0.5 - 1 ug/ml, Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 1 - 2 ug/ml, Immunohistochemistry-Paraffin 1 - 2 ug/ml, CyTOF-ready |
| Applications | Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), CyTOF |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB254540100
![]() |
Novus Biologicals
NBP254540100UG |
100 μg |
Each for $707.00
|
|
|||||
NB254540200
![]() |
Novus Biologicals
NBP254540200UG |
200 μg | N/A | N/A | N/A | ||||
Description
beta-Catenin Monoclonal specifically detects beta-Catenin in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, CyTOF-ready.Specifications
| beta-Catenin | |
| 1 mg/mL | |
| Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), CyTOF | |
| Unconjugated | |
| Mouse | |
| Cancer, Cellular Markers, Extracellular Matrix, Mucosal Immunology, Neuroscience, Stem Cell Signaling Pathway, Wnt Signaling Pathway | |
| PBS with No Preservative | |
| 1499 | |
| Recombinant human full-length beta-Catenin protein fragment | |
| Primary | |
| Store at -20 to -80C. Avoid freeze-thaw cycles. |
| CTNNB1/1509 | |
| Western Blot 0.5 - 1 ug/ml, Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 1 - 2 ug/ml, Immunohistochemistry-Paraffin 1 - 2 ug/ml, CyTOF-ready | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| beta 1 (88kD), beta-catenin, catenin (cadherin-associated protein), beta 1, 88kDa, catenin beta-1, CTNNB, DKFZp686D02253, FLJ25606, FLJ37923 | |
| CTNNB1 | |
| IgG1 κ | |
| Protein A or G purified | |
| 92 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title