Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
beta-Catenin Antibody (OTI12H7), DyLight 550, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270509R
Description
beta-Catenin Monoclonal specifically detects beta-Catenin in Human, Mouse, Rat, Primate, Monkey samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
beta-Catenin | |
Monoclonal | |
DyLight 550 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
CTNNB1 | |
Full length recombinant protein of human beta-Catenin (NP_001895) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat, Primate, Monkey | |
IgG1 |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
OTI12H7 | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
beta 1 (88kD), beta-catenin, catenin (cadherin-associated protein), beta 1, 88kDa, catenin beta-1, CTNNB, DKFZp686D02253, FLJ25606, FLJ37923 | |
Mouse | |
Affinity Purified | |
Cancer, Cellular Markers, Cytoskeleton Markers, Extracellular Matrix, Immune System Diseases, Mucosal Immunology, Neuroscience, Phospho Specific, Signal Transduction, Stem Cell Signaling Pathway, Wnt Signaling Pathway | |
1499.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction