Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

beta-Cyclodextrin 98.0+%, TCI America™
SDP

Catalog No. p-7124675 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
25 g
100 g
500 g

Chemical Identifiers

CAS 7585-39-9
Molecular Formula C42H70O35
Molecular Weight (g/mol) 1134.99
MDL Number MFCD00078139
InChI Key WHGYBXFWUBPSRW-UHFFFAOYNA-N
Synonym cycloheptaamylose, cycloheptapentylose, .beta.-cyclodextrin, caraway, cyclo-hepta-amylose, cycloheptamaltose, .beta.-dextrin, beta-cycloamylose, beta-cyclodextrin, .beta.-cycloamylose
PubChem CID 131707246
IUPAC Name 5,10,15,20,25,30,35-heptakis(hydroxymethyl)-2,4,7,9,12,14,17,19,22,24,27,29,32,34-tetradecaoxaoctacyclo[31.2.2.2³,⁶.2⁸,¹¹.2¹³,¹⁶.2¹⁸,²¹.2²³,²⁶.2²⁸,³¹]nonatetracontane-36,37,38,39,40,41,42,43,44,45,46,47,48,49-tetradecol
SMILES OCC1OC2OC3C(CO)OC(OC4C(CO)OC(OC5C(CO)OC(OC6C(CO)OC(OC7C(CO)OC(OC8C(CO)OC(OC1C(O)C2O)C(O)C8O)C(O)C7O)C(O)C6O)C(O)C5O)C(O)C4O)C(O)C3O

Specifications

Color White-Yellow
Quantity 25 g
Formula Weight 1134.99
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material beta-Cyclodextrin
EINECSNumber (8)-0101
RTECSNumber GU2293000
TSCA Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.