Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ beta-D-glucosidase/beta-D-fucosidase Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5144547
Description
beta-D-glucosidase/beta-D-fucosidase Polyclonal Antibody for Western Blot, ELISA
Bifunctional beta-D-glucosidase/beta-D-fucosidase. Activity towards pNP-beta-D-fucoside is about 80-85% of the activity towards pNP-beta-D-glucoside. Also has slight activity (less than 10%) towards pNP-beta-D-galactoside, and very low activity (less than 1%) towards pNP-beta-D-xyloside. Hydrolyzes laminaribiose, sophorose, cellobiose and gentobiose. Not active against maltose, pNP-alpha-D-glucoside or pNP-beta-L-fucoside.
Specifications
beta-D-glucosidase/beta-D-fucosidase | |
Polyclonal | |
Unconjugated | |
Bifunctional beta-D-glucosidase/beta-D-fucosidase | |
Recombinant Bifidobacterium breve Bifunctional beta-D-glucosidase/beta-D-fucosidase protein (253-460aa). | |
100 μL | |
Primary | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Western Blot | |
4.54 mg/mL | |
PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4 | |
Rabbit | |
Affinity chromatography | |
RUO | |
Bacteria | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction