Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Beta-defensin 106B Antibody, DyLight 350, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP306276UV
Description
Beta-defensin 106B Polyclonal specifically detects Beta-defensin 106B in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Beta-defensin 106B | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
BD6, BD-6, Beta-defensin 6, DEFB106, DEFB106A, DEFB106B, DEFB6, DEFB-6, Defensin, beta 106 | |
Produced in rabbits immunized with E. coli-derived Human Beta-defensin 106B fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
DyLight 350 | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
503841 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction