Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Beta Dystroglycan Antibody (GT9112), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP313642
This item is not returnable.
View return policy
Description
Beta Dystroglycan Monoclonal antibody specifically detects Beta Dystroglycan in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| Beta Dystroglycan | |
| Monoclonal | |
| Unconjugated | |
| A3a, AGRNR, DAG156DAG, dystroglycan, dystroglycan 1 (dystrophin-associated glycoprotein 1), Dystrophin-associated glycoprotein 1 | |
| Recombinant protein encompassing a sequence within the C-terminus region of human Beta Dystroglycan. The exact sequence is proprietary. | |
| 100 μg | |
| Breast Cancer, Cancer | |
| 1605 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| GT9112 | |
| PBS | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction