Learn More
Invitrogen™ beta Dystroglycan Polyclonal Antibody

Description
This antibody is predicted to react with mouse, rabbit, cow and dog based on sequence homology.
Specifications
Specifications
| Antigen | beta Dystroglycan |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.9 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA and 0.02% sodium azide; pH 7.4 |
| Gene | DAG1 |
| Gene Accession No. | Q14118 |
| Gene Alias | 156DAG; A3a; AGRNR; Alpha-DG; Alpha-dystroglycan; Beta-DG; Beta-dystroglycan; D9Wsu13e; DAG; DAG1; Dag-1; DG; Dp427; Dp71; dystroglycan; dystroglycan 1; dystroglycan 1 (dystrophin-associated glycoprotein 1); dystrophin associated glycoprotein 1; Dystrophin-associated glycoprotein 1; MDDGA9; MDDGC7; MDDGC9 |
| Gene Symbols | DAG1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.