Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Beta-endorphin Antibody (B31.15), Alexa Fluor™ 532, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB500414AF532
Description
Beta-endorphin Monoclonal specifically detects Beta-endorphin in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
Beta-endorphin | |
Monoclonal | |
Alexa Fluor 532 | |
50 mM sodium borate with 0.05% sodium azide | |
POMC | |
Human beta Endorphin (full length native protein). | |
Primary | |
The antibody B31.15 reacts with human beta Endorphin, an endogenous opiate derived from ACTH gene. ACTH (Corticotropin; human 39 aa) is synthesized by the anterior pituitary gland and stimulates the adrenal cortex; 6 hormones are derived from one ACTH gene: ACTH, lipotropin, alpha-MSH, beta-MSH, endorphin, and one other. | |
Store at 4°C in the dark. |
Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
B31.15 | |
Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen | |
ACTH, adrenocorticotropic hormone, adrenocorticotropin, alpha-melanocyte-stimulating hormone, alpha-MSH, beta-endorphin, beta-LPH, beta-melanocyte-stimulating hormone, beta-MSH, CLIP, corticotropin-like intermediary peptide, corticotropin-lipotropin, gamma-LPH, gamma-MSH, lipotropin beta, lipotropin gamma, LPH, melanotropin alpha, melanotropin beta, melanotropin gamma, met-enkephalin, MSH, NPP, POC, pro-ACTH-endorphin, proopiomelanocortin, pro-opiomelanocortin, proopiomelanocortin preproprotein | |
Mouse | |
0.1 mL | |
5443 | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction