Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Beta-endorphin Mouse anti-Human, Clone-CLIP/1449, Novus Biologicals™

Mouse Monoclonal Antibody
$617.01
Specifications
| Antigen | Beta-endorphin |
|---|---|
| Clone | CLIP/1449 |
| Concentration | 1 mg/mL |
| Dilution | Flow Cytometry 0.5 - 1 ug/million cells, ELISA 1:100 - 1:2000, Immunocytochemistry/Immunofluorescence 1 - 2 ug/ml, Immunohistochemistry-Paraffin 0.5 - 1.0 ug/ml, CyTOF-ready |
| Applications | Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), CyTOF |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB254324100
![]() |
Novus Biologicals
NBP254324100UG |
100 μg |
Each for $617.01
|
|
|||||
NB254324200
![]() |
Novus Biologicals
NBP254324200UG |
200 μg | N/A | N/A | N/A | ||||
Description
ACTH Monoclonal specifically detects ACTH in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, CyTOF-ready.Specifications
| Beta-endorphin | |
| 1 mg/mL | |
| Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), CyTOF | |
| Unconjugated | |
| RUO | |
| Human | |
| ACTH, adrenocorticotropic hormone, adrenocorticotropin, alpha-melanocyte-stimulating hormone, alpha-MSH, beta-endorphin, beta-LPH, beta-melanocyte-stimulating hormone, beta-MSH, CLIP, corticotropin-like intermediary peptide, corticotropin-lipotropin, gamma-LPH, gamma-MSH, lipotropin beta, lipotropin gamma, LPH, melanotropin alpha, melanotropin beta, melanotropin gamma, met-enkephalin, MSH, NPP, POC, pro-ACTH-endorphin, proopiomelanocortin, pro-opiomelanocortin, proopiomelanocortin preproprotein | |
| POMC | |
| IgG1 | |
| Protein A or G purified |
| CLIP/1449 | |
| Flow Cytometry 0.5 - 1 ug/million cells, ELISA 1:100 - 1:2000, Immunocytochemistry/Immunofluorescence 1 - 2 ug/ml, Immunohistochemistry-Paraffin 0.5 - 1.0 ug/ml, CyTOF-ready | |
| Monoclonal | |
| Mouse | |
| Neuroscience, Nutrient Sensing in the Brain | |
| PBS with No Preservative | |
| 5443 | |
| Synthetic peptide corresponding to aa25-39 of human ACTH (NGAEDESAEAFPLEF) | |
| Primary | |
| Store at -20 to -80C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title