Learn More
Invitrogen™ Beta Endorphin Recombinant Rabbit Monoclonal Antibody (20H4L2)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ 701295
Description
This antibody is predicted to react with mouse, rat, non-human primate and rabbit based on sequence homology. Intact IgG appears on a non-reducing gel as ~150 kDa band and upon reduction generating a ~25 kDa light chain band and a ~50 kDa heavy chain. Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.
Beta endorphin is an opioid neuropeptide that is produced by neurons of the pituitary gland and released into the circulatory system, or produced by the hypothalamus, and released into the brain and spinal cord through nerve fibers. The precursor polypeptide for beta endorphin is pro-opiomelanocortin.
Specifications
Beta Endorphin | |
Recombinant Monoclonal | |
0.5 mg/mL | |
PBS with 0.09% sodium azide | |
P01189 | |
POMC | |
Peptide corresponding to amino acids 242-267 of human Beta Endorphin. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
20H4L2 | |
Unconjugated | |
POMC | |
ACTH; Adrenocorticotropic hormone; adrenocorticotropin; alpha-melanocyte-stimulating hormone; alpha-MSH; Beta-endorphin; beta-LPH; beta-melanocyte-stimulating hormone; beta-MSH; CLIP; Corticotropin; Corticotropin-like intermediary peptide; corticotropin-lipotropin; Gamma-LPH; gamma-MSH; Lipotropin beta; lipotropin gamma; LPH; Melanocyte-stimulating hormone alpha; Melanocyte-stimulating hormone beta; Melanotropin alpha; melanotropin beta; Melanotropin gamma; met-enkephalin; MSH; NPP; opiomelanocortin prepropeptide; OTTHUMP00000119991; OTTHUMP00000200964; POC; POMC; Potential peptide; Precursor of MSH; pro-ACTH-endorphin; proopiomelanocortin; pro-opiomelanocortin; proopiomelanocortin preproprotein | |
Rabbit | |
Protein A | |
RUO | |
5443 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.