Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
beta-Galactosidase-1/GLB1 Antibody (OTI1C9), Novus Biologicals™

Mouse Monoclonal Antibody has been used in 5 publications
Supplier: Novus Biologicals NBP245731
Description
Ensure accurate, reproducible results in Western Blot, Immunohistochemistry
beta-Galactosidase-1/GLB1 Monoclonal specifically detects beta-Galactosidase-1/GLB1 in Human, Canine, Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
beta-Galactosidase-1/GLB1 | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:500, Immunohistochemistry 1:150, Immunohistochemistry-Paraffin 1:150 | |
Acid beta-galactosidase, beta-galactosidase, EBP, EC 3.2.1.23, Elastin receptor 1, elastin receptor 1 (67kD), elastin receptor 1, 67kDa, ELNR1, galactosidase, beta 1, Lactase, MPS4B | |
Mouse | |
72.6 kDa | |
0.1 mL | |
metabolism | |
2720 | |
Store at -20C. Avoid freeze-thaw cycles. | |
IgG2a |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
OTI1C9 | |
Unconjugated | |
PBS (pH 7.3), 1% BSA, 50% Glycerol with 0.02% Sodium Azide | |
GLB1 | |
Full length human recombinant protein of human GLB1(NP_001073279) produced in HEK293T cell. | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Canine, Monkey | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction